Drummond Digital Positive Displacement Microdispenser
Supplier: DRUMMOND SCIENTIFIC COMPANY
Certificates
About this item
Available in five models, this pipettor features a direct digital set and readout for fractional microliters.
- Positive lock prevents accidental volume changes
- Bores fit on 1000 µl unit
- Direct digital set
- Readout for fractional microliters
- Five models available
A positive lock prevents accidental volume changes.
The autoclavable pipettor features an overall volumetric accuracy of ±1%.
Capillary glass bores and polypropylene bores fit on 1000 µl unit.
Ordering information: One pack includes 100 microdispensers.
Frequently Bought Together