- Volumen:0.1 - 10 µl
- Empacado:Bagged
- Referencia:490012-916
- Sterile:-
- Estilo de la punta:Ultra Micro
- Nº de proveedor:104
These pipette tips are made from virgin polypropylene material and are precision molded for an airtight fit using state-of-the-art molds and molding methods to preserve non-wettability in the final product
- Natural
- Metal-free tips: All materials used in the manufacture of these tips are free of trace metals
- Sterile Tips: All sterile tips are sterilized in accordance with AAMI guidelines by gamma irradiation
Autoclavable Tips and Racks: pipette tips and racks are autoclavable at a recommended temperature of 120 °C at 15 pounds of pressure for 15 minutes. Graduated Tips: Provides reference lines to allow a quick check of pipetting procedures by indicating nominal liquid volume in the tip. For use with but not limited to: Pipetman®, Biohit Proline®, Eppendorf®, Jencons Sealpette®, Labpette™, Nichiryo, Oxford-Benchmate®, Rainin, Socorex®, and GeneMate®.
A mold release agent is NOT used in the manufacturing of these pipette tips. A trace metal certificate is available upon request. Certified pipette Tips: Tested by an independent laboratory and certified to be RNAse-, DNAse-, and pyrogen-free. (Non sterilized product is not certified as pyrogen-free.)