Orders LinkContactUsLinkComponent
VWR® Cell Scrapers
VWR® Cell Scrapers
VWR® Cell Scrapers
VWR® Cell Scrapers
Specifically designed to make collecting of cells easier and more effective.
  • Cross-ribbed handle in ABS provides a greater rigidity and ensures a better control while scraping cells
  • Thin, flexible TPE blade prevents damage to cells
  • Individually wrapped
  • Sterilized by electron beam irradiation
  • Non pyrogenic
Available with a choice of blade positions; scraper for use in flasks, or lifter for use in harvesting cells (especially stem cells) in dishes.
  • Catalog No:
  • 10062-904
  • 10062-906
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • ISO 14001/Certificate No.: 04923E00243R0M
  • ISO 14001/Certificate No.: 04923E00243R0M
Product Family Options
  • Description
    Overall Length
    Blade Width
  • Cell Scraper for Flasks
    250 mm
    20 mm
    Case (100UN)
    Case (100UN) - $1,243.52
    $1,243.52
    Case (100UN)
       $1,243.52
      Low Manufacturing Impact
    Specifications
    Cat. No.
    10062-904
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact
  • Cell Scraper for Dishes, Lifter Blade Position
    250 mm
    20 mm
    Case (100UN)
    Case (100UN) - $1,243.52
    $1,243.52
    Case (100UN)
       $1,243.52
      Low Manufacturing Impact
    Specifications
    Cat. No.
    10062-906
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    ISO 14001/Certificate No.: 04923E00243R0M
      Low Manufacturing Impact