Cover Glasses, Rectanglular, Corning®
Catalog # 89239-718
Supplier: Corning
Product available on GSA Advantage®
Specifications
- Numbering:#1
- Descripción:Cover glasses, rectangular
- l×a:50×24 mm
- Grosor:0.12 - 0.16 mm
- Environmentally Preferable:Low Manufacturing Impact
- Certificación ambiental:I-RECs, ISO 14001 - Monterrey
- Referencia:89239-718
- Nº de proveedor:2975-245
Specifications
About this item
Special, optically clear, water-white, borosilicate cover glass is resistant to surface attachment or weathering and will remain clear for extended periods of time.
- Designed for use in general laboratory microscopy
Nonsterile.