ContactUsLinkComponent
VWR® Traceable® Excursion-Trac® Datalogging Refrigerator/Freezer Thermometers
  10048-618
undefined
VWR® Traceable® Excursion-Trac® Datalogging Refrigerator/Freezer Thermometers
  10048-618
  • Resolución:
    0.01°C
  • Descripción:
    VWR® Traceable® Excursion-Trac® Thermometer
  • Referencia:
    10048-618
  • Exactitud:
    ±0.25 °C
  • Sonda:
    1 bottle probe (glycol)
  • Rango:
    –50.0…+70.0 °C (–58…+158 °F)
High/Low Alarm with Time/Date Stamp is an alarm tracking feature which stores in memory each alarm event, detailing when the alarm state occurred, as well as when the unit returned back to within range.
  • Datalogging thermometer with user-defined time intervals
  • Stores up to 10 unique alarm events
  • Meets current CDC requirements for vaccine storage and monitoring
  • Rolling memory structure, maintains most recent 525,600 temperature observations
  • Memory can be cleared on unit after downloading data to USB stick
  • Hassle-free data retrieval
  • Unit can remain in use while downloading and analyzing data
  • Status indicators - low battery, memory full, USB data transfer, and active alarm state
High-accuracy thermometer has a temperature range of –58 to 158°F (–50 to 70°C) and resolution of 0.01°C. It maintains an accuracy of ±0.25°C across the entire temperature range. Ideal for monitoring temperatures in refrigerators, freezers, water baths, heating blocks, and incubators. Bottle probes are sealed in a miniature bottle (1x2½") filled with nontoxic glycol. Solution is deemed GRAS (generally recognized as safe) by the FDA (Food and Drug Administration). Eliminates concerns about incidental contact with food or drinking water. Temperature-buffered bottle sensor eliminates rapid changes when refrigerator door is opened. Ideal for use in monitoring storage conditions for vaccines or other important samples.
High-accuracy thermometer has a temperature range of -58 to 158°F (-50 to 70°C) and resolution of 0.01°C. It maintains an accuracy of ±0.25°C across the entire temperature range. Ideal for monitoring temperatures in refrigerators, freezers, water baths, heating blocks, and incubators. Bottle probes are sealed in a miniature bottle (1x2½") filled with nontoxic glycol. Solution is deemed GRAS (generally recognized as safe) by the FDA (Food and Drug Administration). Eliminates concerns about incidental contact with food or drinking water. Temperature-buffered bottle sensor eliminates rapid changes when refrigerator door is opened. Ideal for use in monitoring storage conditions for vaccines or other important samples.
Data for up to 10 different alarm events can be retrieved with the most current event viewable on the display. A new USB Data Transfer feature allows recorded data (CSV file) to be transferred from thermometer to PC or Mac using a USB flash drive (not included). No software is needed for computer interface. Each thermometer has a unique ID allowing multiple units to be used in the same location. Datalogging features include monitor readings overnight, on weekends, or for any time period with rolling data log. User-defined data-logging interval of one observation/minute to one observation/24 hours. Memory capacity maintains a year of recorded data when using 1 minute intervals and longer when using larger intervals.
 : An individually-numbered Traceable® Certificate is provided which assures accuracy from VWR® ISO 17025 calibration laboratory accredited by A2LA. It indicates traceability to standards provided by NIST (National Institute of Standards and Technology).
 : Includes high-impact, chemical-resistant ABS plastic case (2¾ x 4¼ x ¾"). Weight is 5 ounces. Supplied with stand, Velcro®, magnetic strips, wall mount, Traceable® Certificate, and batteries.