VWR® Digital Mini Incubator
The VWR® digital incubator offers an economical and space saving solution for microbiology or haematology applications.
- Digital controls
- Personal-sized
The temperature can be freely adjusted in increments of 0.1°C up to a maximum of 70°C. Heating elements are incorporated into the housing base and walls to ensure temperature stability and uniformity. The door of the incubator has a transparent plexiglas® window to provide full visibility to samples. The housing and door frame are constructed entirely of epoxy coated mild steel for strength and durability.
: The 230V model is CE marked.
: Warranty–Avantor International warrants that this product will be free from defects in material and workmanship for a period of two (2) years from date of purchase. If a defect is present, Avantor will, at its option, repair, replace, or refund the purchase price of this product at no charge to you, provided it is returned during the warranty period. This warranty does not apply if the product has been damaged by accident, abuse, misuse, or misapplication, or from ordinary wear and tear. For your protection, items being returned must be insured against possible damage or loss. This warranty shall be limited to the replacement of defective products. IT IS EXPRESSLY AGREED THAT THIS WARRANTY WILL BE IN LIEU OF ALL WARRANTIES OF FITNESS AND IN LIEU OF THE WARRANTY OF MERCHANTABILITY.
- Volume:10 L
- Interior Dimensions:230×220×200 mm (9.0×8.6×7.8")
- Exterior Dimensions:285×280×335 mm (11.2×11×13.2")
- Electrical:115 V
- Cat. No.:10055-006
- Temperature uniformity:±0.5 at 37°C
- Temperature range:Ambient 5…70 °C
- Temperature accuracy:±0.5 at 37 °C
- Weight:18 lbs.