ContactUsLinkComponent
SP Bel-Art Processing Trays, Bel-Art Products, a part of SP
SP Bel-Art Processing Trays, Bel-Art Products, a part of SP
Catalog # 70680-050
undefined
SP Bel-Art Processing Trays, Bel-Art Products, a part of SP
Catalog # 70680-050
Supplier Number:  H16108-0000
Specifications
  • Tamaño:
    8 x 10" Print/Film
  • Referencia:
    70680-050
  • Dimensiones interiores:
    33L x 27W x 6H cm (13¹⁄₁₆ x 10¹⁄₁₆ x 2⅜")
  • Nº de proveedor:
    161080000
Specifications
About this item
These trays can be used for staining and blot analysis in biotechnology labs as well as for photo processing.
  • Lightweight
  • Polystyrene plastic construction
  • Cross bars and ribs provide extra rigidity
  • Wide rims
  • Corner spout design
Lightweight polystyrene plastic trays have cross bars and ribs for rigidity. Wide rims and corner spout facilitate pouring.