SP Bel-Art General Purpose Trays, Bel-Art Products, a part of SP
Catalog # 70680-154
Supplier: Bel-Art Products, a Part of SP
Specifications
- Descripción:Tray without Faucet
- Referencia:70680-154
- Dimensiones interiores:41L x 51W x 7.6H cm (16 x 20 x 3”)
- Nº de proveedor:162710000
Specifications
About this item
Molded low-density polyethylene plastic trays are rugged, long-lasting and corrosion-resistant.
- LDPE construction
- Rugged
- Corrosion-resistant
- Heavy-walled design
All trays have heavy walls and wide, top flanges. They will withstand temperatures up to 77ºC (171ºF). Available plain or with F30852-0000 needle-type polyethylene faucet with 12.7 mm (½") bore.