VWR® Cushing Forceps
Stainless steel, 178 mm (7")
Catalog # 89259-952
Supplier: VWR International
Jump to:
Product Details & Documents
VWR® Cushing Forceps are stainless steel with straight, serrated tips.
- Stainless steel construction
- Straight, serrated tips
They feature a corrugated handle and a guide pin.
Especificaciones
- Length:178 mm (7")
- Material:Stainless steel