Orders LinkContactUsLinkComponent
VWR® Dissecting Forceps with Guide Pin
VWR® Dissecting Forceps with Guide Pin
Dissecting Forceps with Straight Tips, Stainless steel, 114 mm (4½")
  89259-944
VWR® Dissecting Forceps with Guide Pin
  89259-944
VWR® Dissecting Forceps with Guide Pin
VWR® Dissecting Forceps with Guide Pin
Dissecting Forceps with Straight Tips, Stainless steel, 114 mm (4½")
$63.59
Each (1UN) Paquete de 12 (1UN)
VWR® Dissecting Forceps are stainless steel with fine-point serrated tips and a guide pin.
  • Stainless steel construction
  • Fine-pointed serrated tips
Dissecting forceps are used for a variety of applications and are available in different lengths.
Documents
  • Description:
    Dissecting Forceps with Straight Tips
  • Length:
    114 mm (4½")
  • Material:
    Stainless steel