VWR® Reverse Action Forceps
Stainless steel, 127 mm (5")
Supplier: VWR International
Jump to:
Product Details & Documents
VWR® Reverse Action Forceps open with finger pressure.
- Stainless steel construction
- Finger-pressure open action
- Sharp, smooth points
They are stainless steel with sharp, smooth points.
Documents
Certificates
Buscar por número de loteEspecificaciones
- Length:127 mm (5")
- Material:Stainless steel