Orders LinkContactUsLinkComponent
Camel Hair Brushes, Electron Microscopy Sciences
Camel Hair Brushes, Electron Microscopy Sciences
Camel Hair Round Brush, Camel hair, 1 1/2'', 19 mm (3/4"), Wood, 9"
Catalog # 102095-920
Número de proveedor #:  65575-01
Camel Hair Brushes, Electron Microscopy Sciences
Catalog # 102095-920
Camel Hair Brushes, Electron Microscopy Sciences
Camel Hair Brushes, Electron Microscopy Sciences
Camel Hair Round Brush, Camel hair, 1 1/2'', 19 mm (3/4"), Wood, 9"
Price
$335.16
Log in Para disponibilidad
Each (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Camel hair brushes are used for dusting and cleaning embedding center, balances, and sensitive instruments. Camel hair is mounted on wooden handle with metal ferrule.
  • Round, camel hair portion has full body and slightly pointed tip. Brush size 3/4" diameter x 11/2" long
  • Paint brush style 1, camel hair portion is 1" long x 1" wide
Documents
Especificaciones
  • Bristle Material:
    Camel hair
  • Brush head Ø:
    19 mm (3/4")
  • Handle Material:
    Wood
  • Description:
    Camel Hair Round Brush
  • Total length:
    9"
  • Brush Head Length:
    1 1/2''
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 