Feather® Sterile Micro Scalpels; Stainless Steel, Electron Microscopy Sciences
Catalog # 100491-034
Supplier: ELECTRON MICROSCOPY SCIENCES
Minority or Woman-Owned Business Enterprise
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Descripción:Micro-Scalpel, Feather, 15°
- Referencia:100491-034
- Nº de proveedor:72045-15
Specifications
About this item
Micro precision, ultra sharp points.
- Sterilized by gamma irradiation
- Disposable
- Sure-grip aluminum handle-color coded for easy angle identification
- Comes in a carbonate plastic case
- Available in 3 angle styles: 15°, 30° and 45°