Orders LinkContactUsLinkComponent
Loctite Adhesive, Electron Microscopy Sciences
Loctite Prism 4011, 20 g
  102098-054
 :  72573-11
Loctite Adhesive, Electron Microscopy Sciences
  102098-054
 :  
Loctite Adhesive, Electron Microscopy Sciences
Loctite Adhesive, Electron Microscopy Sciences
Loctite Prism 4011, 20 g
 :  
$2,105.08
Each (1UN)
A variety of Loctite adhesives are available that will bond a range of materials.
  • 409™ Super Bonder®
  • 454™ Prism®
  • 861™ Prism®
  • 349™ Impruv®
  • 4011™ Prism®
The 409™ Super Bonder® is a general-purpose gel adhesive, clear, gap fills 0.010”; surface insensitive Ethyl, tensile shear strength 3,200 psi, and temperature range: –65°F to 180°F
The 454™ Prism® is for use with porous surfaces, clear, gap fills 0.010”; surface insensitive Ethyl, gel type, tensile shear strength 3200 psi, temperature range: –65°F to 180°F. Cure speed: fixture: 15 seconds, full: 24 hours.
The 861™ Prism® is for use with flexible surfaces, clear, gap fill 0.008”; surface flexible Alkyl, viscosity 4,000 cP, tensile shear strength 2465 psi, temperature range: –65°F to 212°F. Cure speed: fixture: 25 seconds, full: 24 hours.
The 349™ Impruv® is for bonding glass/metal; appearance: clear/straw, cure type: UV, viscosity: 9,500cP, shore hardness: D70, temperature range: –65°F to 266°F.
Cure speed: fixture: 75 seconds, full: 24 hours.
  • Description:
    Loctite Prism 4011
  • Size:
    20 g
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.