Dust Cover
Dust cover for large microscope, 35.5 cm x 43 cm tall
Catalog # 470014-572
Supplier: MOTIC HONG KONG LIMITED
Número de proveedor #: MI0080
Jump to:
Product Details & Documents
Fits most full-size microscopes.
Especificaciones
- Description:Dust cover for large microscope, 35.5 cm x 43 cm tall