ContactUsLinkComponent
SP Bel-Art Polypropylene Forceps, Bel-Art Products, a part of SP
  70690-142
undefined
SP Bel-Art Polypropylene Forceps, Bel-Art Products, a part of SP
  70690-142
 :  F37920-0000
  • Longitud:
    12.7 cm (5”)
  • Referencia:
    70690-142
  • Nº de proveedor:
    379200000
These polypropylene plastic forceps have serrated tips to ensure a firm grasp.
  • Polypropylene plastic construction
  • Strong and durable
  • Autoclavable
  • Serrated tips for a firm grip
Strong and durable, they are steam autoclavable up to 121ºC (250ºF).