ContactUsLinkComponent
Iron Test Method: Photometric 0.005 - 5.00 mg/L Fe Spectroquant®
Iron Test Method: Photometric 0.005 - 5.00 mg/L Fe Spectroquant®
Catalog # EM1.14761.0002
CAS Number:  
Iron Test Method: Photometric 0.005 - 5.00 mg/L Fe Spectroquant®
Catalog # EM1.14761.0002
Supplier Number:  EM1.14761.0002
CAS Number:  
Specifications
  • Tipo de env.:
    Cardboard Box(es)
  • Tamaño:
    1 pc
  • Referencia:
    EM1.14761.0002
  • Nº de proveedor:
    1.14761.0002