ContactUsLinkComponent
Residual Hardness Cell Test Method: photometric 0.50 - 5.00 mg/l Ca 0.070 - 0.700 °d 0.087 - 0.874 °e 0.12 - 1.25 °f 0.70 - 7.00 CaO 1.2 - 12.5 mg/l CaCO3 Spectroquant®
Residual Hardness Cell Test Method: photometric 0.50 - 5.00 mg/l Ca 0.070 - 0.700 °d 0.087 - 0.874 °e 0.12 - 1.25 °f 0.70 - 7.00 CaO 1.2 - 12.5 mg/l CaCO3 Spectroquant®
Catalog # EM1.14683.0001
CAS Number:  
Residual Hardness Cell Test Method: photometric 0.50 - 5.00 mg/l Ca 0.070 - 0.700 °d 0.087 - 0.874 °e 0.12 - 1.25 °f 0.70 - 7.00 CaO 1.2 - 12.5 mg/l CaCO3 Spectroquant®
Catalog # EM1.14683.0001
Supplier Number:  EM1.14683.0001
CAS Number:  
Specifications
  • Tipo de env.:
    EPS-Moulding
  • Tamaño:
    1 pc
  • Referencia:
    EM1.14683.0001
  • Nº de proveedor:
    1.14683.0001