ContactUsLinkComponent
Sulfate Cell Test Method: photometric 5 to 250 mg/L SO₄²⁻ Spectroquant®, MilliporeSigma
Sulfate Cell Test Method: photometric 5 to 250 mg/L SO₄²⁻ Spectroquant®, MilliporeSigma
Catalog # EM1.14548.0001
CAS Number:  
Sulfate Cell Test Method: photometric 5 to 250 mg/L SO₄²⁻ Spectroquant®, MilliporeSigma
Catalog # EM1.14548.0001
Supplier Number:  1.14548.0001
CAS Number:  
Specifications
  • Tipo de env.:
    EPS-Moulding
  • Tamaño:
    1 pc
  • Referencia:
    EM1.14548.0001
  • Nº de proveedor:
    1.14548.0001