ContactUsLinkComponent
Ammonium Cell Tests, Method: photometric, Spectroquant®, Supelco®
Ammonium Cell Tests, Method: photometric, Spectroquant®, Supelco®
Catalog # EM1.14739.0001
CAS Number:  
Ammonium Cell Tests, Method: photometric, Spectroquant®, Supelco®
Catalog # EM1.14739.0001
Supplier Number:  1147390001
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Método químico:
    Indophenol blue
  • Descripción:
    Ammonium cell test
  • Tamaño:
    1 pc
  • Environmentally Preferable:
  • Referencia:
    EM1.14739.0001
  • Nº de proveedor:
    1.14739.0001
  • No. of Tests:
    25
  • Para la unidad:
    Prove, Nova 30/60, Multy, Move
Specifications
About this item
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments
  • Very user friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells for most the test parameters
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times.
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).