Orders LinkContactUsLinkComponent
Test Kits, COD Cell Test Method: Photometric Spectroquant®, Supelco®
Test Kits, COD Cell Test Method: Photometric Spectroquant®, Supelco®
COD cell test*7-1, 25 Tests, Chromosulphuric oxidation, Prove, Nova 30/60, Multy, Move, 15 - 300 mg/L
Catalog # EM1.14895.0001
Supplier:   Avantor
Test Kits, COD Cell Test Method: Photometric Spectroquant®, Supelco®
Catalog # EM1.14895.0001
Supplier:  Avantor
CAS Number:  
Test Kits, COD Cell Test Method: Photometric Spectroquant®, Supelco®
COD cell test*7-1, 25 Tests, Chromosulphuric oxidation, Prove, Nova 30/60, Multy, Move, 15 - 300 mg/L
CAS Number:  
Price
$4,099.59
Log in Para disponibilidad
Each (1UN) Case de 8 (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments. Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
  • Very user-friendly, validated methods
  • All the necessary reagents for the analysis are contained in the cells
  • Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
  • The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set up times.
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
*7 This method is officially recognised by the USEPA as an alternative method for the investigation of the following:
*7-1 wastewater
*7-2 drinking water
*7-3 respectively drinking water and waste water
*8 COD cell tests fully comply with ISO 15705
Especificaciones
Especificaciones
  • Chemical method:
    Chromosulphuric oxidation
  • Description:
    COD cell test*<sup>7-1</sup>
  • Size:
    25 Tests
  • Range:
    15 - 300 mg/L
  • For Unit:
    Prove, Nova 30/60, Multy, Move
Especificaciones
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 