ContactUsLinkComponent
Total Hardness Cell Test Method: photometric 5 - 215 mg/l Ca 0.7 - 30.1 °d 0.9 - 37.6 °e 1.2 - 53.7 °f 7 - 301 mg/l CaO 12 - 537 mg/l CaCO3 Spectroquant®
Total Hardness Cell Test Method: photometric 5 - 215 mg/l Ca 0.7 - 30.1 °d 0.9 - 37.6 °e 1.2 - 53.7 °f 7 - 301 mg/l CaO 12 - 537 mg/l CaCO3 Spectroquant®
Catalog # EM1.00961.0001
CAS Number:  
Total Hardness Cell Test Method: photometric 5 - 215 mg/l Ca 0.7 - 30.1 °d 0.9 - 37.6 °e 1.2 - 53.7 °f 7 - 301 mg/l CaO 12 - 537 mg/l CaCO3 Spectroquant®
Catalog # EM1.00961.0001
Supplier Number:  1.00961.0001
CAS Number:  
Specifications
  • Tipo de env.:
    EPS-Moulding
  • Tamaño:
    1 pc
  • Referencia:
    EM1.00961.0001
  • Nº de proveedor:
    1.00961.0001