Spectroquant® Nitrate Cell Tests, Photometric Method, MilliporeSigma
All Spectroquant® test kits can be used with the Prove range of spectrophotometers and Nova 60/60A instruments
- Very user friendly, validated methods
- All the necessary reagents for the analysis are contained in the cells for most the test parameters
- Packaging design protects the cells from fluctuation of temperature and moisture during storage and transport
- The barcode on the cells ensures the correct method of analysis is chosen - eliminating mistakes and reducing set-up times.
Note: The range quoted in the table below refers to the Prove and NOVA instruments. For some test kits the measuring range available for the Spectroquant® Multy and Move 100 instruments may be reduced.
Tests can be used not only on photometers and spectrophotometer from Merck, but also on photometers and spectrophotometers from other suppliers (programming details available on request).
Documents
MilliporeSigma Industrial Applications MilliporeSigma Pharma Brochure
Documentación del producto
VWR GHS Compliance Letter (US)
Detalles adicionales del producto
- Chemical method:Nitrospectral
- Pack type:EPS-Moulding
- Size:1 pc
- Range:0.5 - 18.0 mg/L NO3-N; 2.2 - 79.7 mg/l NO3-
- No. of Tests:25
- For Unit:Prove, Nova 30/60, Multy, Move
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.