ContactUsLinkComponent
Spectroquant® Nitrogen (total) Cell Test Method: photometric, DMP 0.5 - 15.0 mg/L N
Spectroquant® Nitrogen (total) Cell Test Method: photometric, DMP 0.5 - 15.0 mg/L N
Catalog # EM1.00613.0001
CAS Number:  
Spectroquant® Nitrogen (total) Cell Test Method: photometric, DMP 0.5 - 15.0 mg/L N
Catalog # EM1.00613.0001
Supplier Number:  1.00613.0001
CAS Number:  
Specifications
  • Tipo de env.:
    EPS-Moulding
  • Tamaño:
    1pc
  • Referencia:
    EM1.00613.0001
  • Nº de proveedor:
    1.00613.0001