Pluro Stoppers, Kimble Chase
Catalog # 46600-788
Supplier: DWK Life Sciences (KIMBLE)
Specifications
- Descripción:Set of 7 Neoprene Rubber Pluro Stoppers
- Referencia:46600-788
- Nº de proveedor:852050-0070
Specifications
About this item
This autoclavable neoprene rubber adapter set consists of seven cones designed to fit buchner and fritted glass funnels
Set includes one of each sizes 1 through 7.
Can be used singly, or nested with adjacent sizes. Eliminates boring of special size holes in rubber stoppers and difficulties of insertion and removal.