SP Bel-Art Poxygrid® Baskets, Bel-Art Products, a part of SP
Catálogo # 13195-150
Proveedor: Bel-Art Products, a Part of SP
Especificaciones
- Dimensiones:63L x 36W x 15H cm (24¾ x 14 x 6")
- Referencia:13195-150
- Peso al enviarse:3.0 kg (6.5lb)
- Nº de proveedor:167590000
Especificaciones
Acerca de este artículo
Epoxy-coated steel wire baskets hold up under the most severe laboratory conditions and are handy for transporting and batch dipping glassware.
- Epoxy-coated steel wire construction
- Corrosion- and chemical-resistant
- Autoclavable
They resist rust, corrosion, solvents, salts, weak acids and weak alkalis, will withstand a temperature range of -223ºC (-369ºF) to 121ºC (250ºF) and can be autoclaved.