Orders LinkContactUsLinkComponent
Minncare® Test Strips, Mar Cor Purification
Minncare® Test Strips, Mar Cor Purification
Minncare® Test Strips, Mar Cor Purification
Certificates
Minncare® Test Strips, Mar Cor Purification
2 Options Available Below
Product Details & Documents
Minncare 1% Test Strips verify adequate concentration of a 1% Minncare solution during sanitization of your high purity water system
Minncare Residual Test Strips are easy to use and are conveniently packaged 100 test strips per container. Each strip has a “reaction zone” pad located at one end of the strip. When stored and used properly, the test strip pad can indicate residual Minncare levels as low as 1 part per million (ppm). The reaction zones are set to provide six different reading levels at 100, 30, 10, 3, 1 and 0 (zero) parts per million.
These convenient dip-and-read strips can be used at any test port in your system to provide an accurate pass/fail measurement of Minncare solution.
Product Family Options
Información del producto
  • Availability
    Price
  • 50930-070
    Minncare 1% Test Strips
    Minncare 1% Test Strips
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $1,396.05
    $1,396.05
    Paquete de 100
       $1,396.05
    Supplier Number: 78339000
  • 50930-068
    Minncare Residual Test Strips
    Minncare Residual Test Strips
    Log in Para disponibilidad
    Log in Para disponibilidad
    Paquete de 100
    Paquete de 100 - $1,425.70
    $1,425.70
    Paquete de 100
       $1,425.70
    Supplier Number: 78338000