Amplitube™ PCR Reaction Tubes, 0.2 ml, Simport Scientific
Catálogo # 89429-020
Proveedor: SIMPORT SCIENTIFIC INC.
Especificaciones
- Cap Type:Flat cap
- Capacidad:0.2 ml
- Color:Red
- Descripción:PCR Reaction Tube and Cap with Integral Shield
- Referencia:89429-020
- Nº de proveedor:T325-2R
Especificaciones
Acerca de este artículo
Amplitube™ PCR Reaction tubes are made of transparent superior grade polypropylene for improved viewing of the sample contents.
- Attached hinged caps are either dome or flat-topped
- Caps provide positive sealing during thermal cycling and prevent evaporation
- Caps can be easily opened and closed with one hand
- Autoclavable
Tubes are available with or without an integral shield that prevents contamination with surface of lid. A frosted writing surface is provided for sample identification.
Ultra-thin wall design ensures rapid thermal transfer and significant reduction in cycle and PCR reaction time.
Rnase-, Dnase-, Pyrogen-, and DNA-free.
Información de pedido: Available in colorless or four non-cytotoxic colors.
Embalaje: Packaged in tamper-proof, resealable safety-lock bags.