SP Bel-Art Conway Diffusion Cell, Bel-Art Products, a part of SP
Catalog # 70690-140
Supplier: Bel-Art Products, a Part of SP
Specifications
- Ext.Ø:83 mm
- Descripción:Conway Diffusion Cell
- Referencia:70690-140
- Nº de proveedor:409410000
Specifications
About this item
Designed for determining carbon dioxide, ammonia, or alcohol in blood or serum by diffusion method.
- White, molded polypropylene construction
- Natural, clear polypropylene cover for extra visibility
- Autoclavable
Smooth, white, molded polypropylene with a natural, clear polypropylene cover
The Diffusion Cell has a moat depth of 4 mm. The chamber depth is 10 mm, with a 60 mm diameter. The center depth is 4 mm with a 35 mm diameter.
The cover has a lifting knob and fits the annular moat of the cell to seal off the chamber. Steam autoclavable at 121°C (250°F).