ContactUsLinkComponent
SP Bel-Art Micro Funnels, Bel-Art Products, a part of SP
SP Bel-Art Micro Funnels, Bel-Art Products, a part of SP
Catalog # 13197-128
undefined
SP Bel-Art Micro Funnels, Bel-Art Products, a part of SP
Catalog # 13197-128
Supplier Number:  F14685-0035
Specifications
  • Capacidad:
    8.1 ml (0.27 oz.)
  • Referencia:
    13197-128
  • D.I. superior:
    35 mm
  • Diámetro del vástago:
    8.0 mm
  • Nº de proveedor:
    F146850035
Specifications
About this item
Prefect micro funnels for micro work, filling burettes, manometers, and small bottles.
  • Polypropylene construction
  • Suitable for micro work
  • Autoclavable
Steam autoclavable at 121 °C (250 °F).