ContactUsLinkComponent
Unjacketed Benchtop Reactor, All-In-One, Ace Glass Incorporated
Unjacketed Benchtop Reactor, All-In-One, Ace Glass Incorporated
Catalog # 89055-512
Supplier:  ACE GLASS, INC.
Product available on GSA Advantage®
Small Business Enterprise
undefined
Unjacketed Benchtop Reactor, All-In-One, Ace Glass Incorporated
Catalog # 89055-512
Supplier:  ACE GLASS, INC.
Supplier Number:  6542-12
Product available on GSA Advantage®
Small Business Enterprise
Specifications
  • Capacidad:
    1 L
  • Referencia:
    89055-512
  • Nº de proveedor:
    6542-12
Specifications
About this item
A complete, general purpose benchtop reactor.
  • All wetted surfaces are glass or PTFE
  • A sturdy stand and all necessary clamps and clips are included
Self-supported with a 20" x 15" footprint, 33" high. Reactor features a 100 mm(4") head with (4) 24/40 joints, an allihn condenser, a pressure equalizing funnel, overhead glass stirring assembly with variable speed motor and controller, heating mantle with type J PTFE covered probe and controller.
If working with pressure, We recommend the 89054-494 safety manifold.