ContactUsLinkComponent
SP Bel-Art Mix and Match Quick Disconnects, Bel-Art Products, a part of SP
  13197-344
undefined
SP Bel-Art Mix and Match Quick Disconnects, Bel-Art Products, a part of SP
  13197-344
 :  H19727-0000
  • Tubo Ø int.:
    3 - 6 mm (⅛ - ¼”)
  • Referencia:
    13197-344
  • Nº de proveedor:
    197270000
Full range of sizes to fit tubing from 3 to 12 mm (⅛ to ½") I.D.
  • Polyethylene plastic construction
  • Useful in connecting vacuum lines and other tubing assemblies
  • Interchangeable sizes make reducing combinations
Consists of two barbed polyethylene plastic connectors which assemble tightly together with a male-female center taper. Very useful in connecting and disconnecting vacuum lines and other tubing assemblies. All sizes are interchangeable with others to make reducing combinations. Operates at temperatures up to 80°C (176°F).