Orders LinkContactUsLinkComponent
VWR® Stackable Freezer Boxes
VWR® Stackable Freezer Boxes
VWR® Stackable Freezer Boxes
VWR® Stackable Freezer Boxes
VWR® Stackable Freezer Boxes are suitable to store in temperatures as low as –86 °C.
  • Made with PP material
  • Base made with color options natural, blue, green, purple, and orange
  • Lid has grid option for coding
Boxes are stackable. Designed for 50, 81, or 100 places to store both internal and external threaded vials (1 to 2 ml).
Slot width: 12.5 mm (±0.3 mm)
Documents
Product Family Options
  • No. of Holes
    For Vials
    Color
    Dimensions
  • 50
    1.8 ml
    Mixed
    71.6W×139.8L×56.7H mm
    Paquete de 5
    Paquete de 5 - $341.28
    $341.28
    Case de 10 (5UN)
    Case de 10 (5UN) - $371.99
    $371.99
    Paquete de 5
       $341.28
    Case de 10 (5UN)
       $371.99
  • 81
    1.8 ml
    Mixed
    126.5W×126.5L×56.7H mm
    Paquete de 5
    Paquete de 5 - $490.52
    $490.52
    Case de 10 (5UN)
    Case de 10 (5UN) - $534.67
    $534.67
    Paquete de 5
       $490.52
    Case de 10 (5UN)
       $534.67
  • 81
    1.8 ml
    Natural
    126.5W×126.5L×56.7H mm
    Paquete de 5
    Paquete de 5 - $490.52
    $490.52
    Case de 10 (5UN)
    Case de 10 (5UN) - $534.67
    $534.67
    Paquete de 5
       $490.52
    Case de 10 (5UN)
       $534.67
  • 100
    1.8 ml
    Mixed
    140.6W×140.6L×57.2H mm
    Paquete de 5
    Paquete de 5 - $558.43
    $558.43
    Case de 10 (5UN)
    Case de 10 (5UN) - $608.69
    $608.69
    Paquete de 5
       $558.43
    Case de 10 (5UN)
       $608.69