ContactUsLinkComponent
SP Bel-Art Chemical Containers, Bel-Art Products, a part of SP
SP Bel-Art Chemical Containers, Bel-Art Products, a part of SP
Catalog # 70690-008
undefined
SP Bel-Art Chemical Containers, Bel-Art Products, a part of SP
Catalog # 70690-008
Supplier Number:  H17874-0000
Specifications
  • Tamaño de la tapa:
    54 mm
  • Ext.Ø:
    52 mm
  • Capacidad:
    90 ml
  • Altura:
    69 mm
  • Referencia:
    70690-008
  • Nº de proveedor:
    H178740000
Specifications
About this item
Containers are great for storing small quantities of liquids or solids.
  • Heavy-walled design
  • Chemically resistant
  • Easy to clean
  • Leakproof inner seal
These heavy-walled, injection molded polyethylene jars have even been used to transport dust samples from the moon. Easy-to-clean with a screw cap design featuring a leak-proof inner seal.