Orders LinkContactUsLinkComponent
Analytical Funnels, Globe Scientific
Analytical Funnels, Globe Scientific
PP, 5 mm, 37 mm, 35 mm
Catalog # 10027-882
Número de proveedor #:  600146
Analytical Funnels, Globe Scientific
Catalog # 10027-882
Analytical Funnels, Globe Scientific
Analytical Funnels, Globe Scientific
PP, 5 mm, 37 mm, 35 mm
Price
$1,482.49
Log in Para disponibilidad
Case (20UN)
Product Details & Documents
These heavy duty polypropylene (PP) analytical funnels are perfect for use in analytical chemistry applications
  • Heavy duty polypropylene (PP)
  • Autoclavable
  • Food safe
  • Air lock prevention
These analytical funnels have internal and external ribbing which helps to prevent air lock. They are angled at exactly 60° for rapid filtration.
Available in 9 different sizes, these analytical funnels accommodate standard filter paper sizes.
They feature long, narrow stems, which make them ideal for use with liquids.
Especificaciones
  • Material:
    PP
  • Top Diameter:
    35 mm
  • Stem Length:
    37 mm
  • Stem Diameter:
    5 mm