Pipe and Cylinder Brushes, Perfex
Excellent for cleaning valves, outlets, tanks, bottles, pitchers, pots, vats, drains, etc.
- Non-conductive and will not absorb bacteria, liquids or odors
- Unaffected by water, grease, petroleum products, detergents, sanitizers and solvents
Moderately stiff polyester fibers will not collapse and quickly rinse odor free.
Frequently Bought Together