SP Bel-Art Buchner Funnels, Single Piece, Bel-Art Products, a part of SP
Catalog # 70690-258
Supplier: Bel-Art Products, a Part of SP
Specifications
- Capacidad:1000 ml
- Referencia:70690-258
- Borde a placa:94 mm
- Plate Diameter:122 mm
- Nº de proveedor:146110000
Specifications
About this item
One-piece HDPE plastic funnels with sealed-in, polyethylene filter plate of coarse porosity (90-130 microns) offer good retention and fast flow under vacuum.
- One-piece design
- HDPE funnel construction
- Polyethylene filter plate
- Good retention and fast flow
Useable with or without filter paper. Use below 52ºC (125ºF). Use with Vac-Ring Filter Seal (H38959-0000) for quick filter setup.