SP Bel-Art Buchner Funnels, Two Piece, Bel-Art Products, a part of SP
30306-430
- Capacidad:875 ml
- Referencia:30306-430
- Se ajusta al diá. del papel:110 mm
- Borde a placa:67 mm
- Nº de proveedor:H14604-0000
Easy-to-clean two-piece, polypropylene plastic Buchner funnels include a molded-in perforated plate.
- Two-piece design
- Polypropylene construction
- Molded-in perforated plate
- Lightweight
- Autoclavable
Lightweight, but not top-heavy. Steam autoclavable at 121ºC (250ºF). Use with Vac-Ring Filter Seal (H38959-0000) for quick filter setup.