Orders LinkContactUsLinkComponent
J.T.Baker® BAKER ANALYZED®, Buffer Solution, pH 7, Color Coded
J.T.Baker® BAKER ANALYZED®, Buffer Solution, pH 7, Color Coded
± 0.04, 25 °C, PE bottle, 500 ml, 7.00, Yellow
Catalog # JT5656-1
Número de proveedor #:  5656-01
J.T.Baker® BAKER ANALYZED®, Buffer Solution, pH 7, Color Coded
Catalog # JT5656-1
CAS Number:  
MOC:  No regulado, con informaciónCertificates
J.T.Baker® BAKER ANALYZED®, Buffer Solution, pH 7, Color Coded
J.T.Baker® BAKER ANALYZED®, Buffer Solution, pH 7, Color Coded
± 0.04, 25 °C, PE bottle, 500 ml, 7.00, Yellow
CAS Number:  
MOC:  No regulado, con información
Price
$442.09
Log in Para disponibilidad
Each(1/Itm/Bx) Case de 12 (1/ Itm/Bx)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This BAKER ANALYZED® Reagent is a phosphate buffer solution for laboratory, research, and/or manufacturing uses.
  • Contains no presevative
Each lot is compared to NIST Standard Reference Buffer.
Documents
Certificates
Buscar por número de lote
Especificaciones
Especificaciones
  • Pack type:
    PE bottle
  • Color:
    Yellow
  • Size:
    500 ml
  • Reference Temperature:
    25 °C
  • pH:
    7.00
  • pH Accuracy:
    ± 0.04
Especificaciones
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 