Orders LinkContactUsLinkComponent
BD BBL™ Eosin Methylene Blue Agar, BD Diagnostics
BD BBL™ Eosin Methylene Blue Agar, BD Diagnostics
BD BBL™ Eosin Methylene Blue Agar, BD Diagnostics
This dehydrated culture media is used for isolating and differentiating lactose-fermenting from lactose-nonfermenting gram-negative enteric bacilli.
Product Family Options
  • Description
    Size
  • BD BBL™ Eosin Methylene Blue Agar, Levine
    500 g
    Each (500g)
    Each (500g) - $5,290.00
    $5,290.00
    Each (500g)
       $5,290.00
     : 211221
  • BD BBL™ Eosin Methylene Blue Agar, Modified (Holt-Harris & Teague)
    1 lb.
    Each (1lb)
    Each (1lb) - $5,950.00
    $5,950.00
    Each (1lb)
       $5,950.00
     : 211215