SP Bel-Art Spinbox™ Magnetic Stirring Bar Assortments, Bel-Art Products, a part of SP
Catalog # 76001-836
Supplier: Bel-Art Products, a Part of SP
Specifications
- Revestimiento:PTFE
- Descripción:Assorted Octagon Stirring Bars, Standard
- Referencia:76001-836
- Forma:Octagon
Specifications
About this item
These stirbar assortments come in a convenient storage box and provide instant availability of these highly popular Teflon® PTFE encapsulated Alnico magnets.
- PTFE low-friction coating
- Durable and inert
- Autoclavable
Choose from Octagon, Cylindrical, Polygon, Micro, Egg-Shaped or Spinplus® shaped assortments.
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.