SP Bel-Art Circulus™ Magnetic Stirring Bars, Bel-Art Products, a part of SP
Catalog # 30617-696
Supplier: Bel-Art Products, a Part of SP
Specifications
- Descripción:White Disk
- Longitud:32 mm (1¼")
- Referencia:30617-696
- Nº de proveedor:371700000
Specifications
About this item
Circulus™ Magnetic Stirring Bars offer fast mixing action and strong turbulence as well as greater stability with less drifting at higher speeds.
- Ideal for containers with round or convex bottoms
- Fast mixing action
- Solid PTFE end disks
- Autoclavable
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.