SP Bel-Art Magnetic Stirring Bar Retriever, Bel-Art Products, a part of SP
Catalog # 47750-232
Supplier: Bel-Art Products, a Part of SP
Specifications
- Descripción:Magnetic Stirring Bar Retriever
- Referencia:47750-232
- Nº de proveedor:377750000
Specifications
About this item
This retriever is able to remove a magnetic stirring bar without coming into contact with the contents of the vessel.
- HDPE construction
- Retriever eliminates possibility of sample contamination
- No need for autoclaving after use
- Long, ribbed handle for secure grip
It eliminates sample contamination and does not need to be cleaned or autoclaved after use. The retriever is held in contact with the vessel where the magnetic stirring bar rests. It is then moved upward with the stirring bar following on the inside of the vessel to a point where it can be grasped. The magnet is located in the 25 x 70 mm (1 x 23/4") rectangular section, and a long, ribbed handle provides a secure grip. Made of high-density polyethylene. Overall length is 20 cm (7.7").