SP Bel-Art Spinbar® Polygon, Magnetic Stirring Bars without Pivot Ring, Bel-Art Products, a part of SP
Proveedor: Bel-Art Products, a Part of SP
Certificados
Acerca de este artículo
These stirbars have polygon multifaceted surfaces that add turbulence relative to similar smooth size cylindrical bars.
- PTFE low friction coating
- Durable and inert
- Autoclave safe
Manufactured in a registered ISO 9001:2008 facility, as verified by SGS certification.
Información relacionada
Certificados
Buscar por número de loteAT-Bel-Art Stirring Bar Selection and Use Guide.pdf
Hoja de datos
Frequently Bought Together