ContactUsLinkComponent
Urine Collection Cups, Globe Scientific
  89508-716
undefined
Urine Collection Cups, Globe Scientific
  89508-716
 :  5915
  • Esterilidad:
    Non sterile
  • Capacidad:
    120 ml (4 oz.)
  • Referencia:
    89508-716
  • Empaquetado:
    Bulk
  • Cap Color:
    Red
  • Nº de proveedor:
    5915
These popular urine collection cups feature a quarter turn screw cap which is designed to assist in the prevention of carpal tunnel syndrome.
  • Polypropyelene (PP)
  • Cap Material: Polyethylene (PE)
  • Container Dimensions: 56.8×72.5 mm
  • Cap Dimensions: 61 mm
Containers are graduated to 100 ml and feature a frosted writing area.
Sterile containers are individually wrapped and feature a trilingual patient ID label (English, Spanish, and French).
The lid opens and closes with one smooth movement, which minimizes the usage of the operator's hand and lower arm area, and reduces the stress placed on the wrist.
 : A temperature strip is attached to collection cup 76293-172.