Select® Cover Glasses, Propper Manufacturing
48300-484
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
- Descripción:Glass cover slips, square
- l×a:22×22 mm
- Grosor:0.13 - 0.17 mm
- Referencia:48300-484
- Nº de proveedor:141102
Propper Select® cover glasses are made of high quality clear, non-fogging, non-corrosive glass.
- Ready to use
- For use with all levels of microscopy
- Highly resistant to breakage
Propper Cover Glass has parallel surfaces and uniform homogeneity and allow for uniform wetting. Each cover glass is cleaned, washed and ready to use for all levels of microscopy. Available in thickness from 0.13 to 0.25 mm.