PYREX® Disposable Glass Conical Centrifuge Tubes, Ungraduated, without Screw Cap, Corning
Supplier: Corning
Certificates
About this item
Corning disposable glass centrifuge tubes are manufactured from PYREX® Type I borosilicate glass.
These tubes feature more consistent uniform wall thickness, a well-formed conical tapered tip and durable tip radius. The optimally shaped tip design allows for easy removal of samples following centrifugation. The tubes are designed to withstand centrifugation up to 3000 RCF and are packed in convenient, easy to use packs, which are shrink sealed for protection during shipment.
Ordering information: All caps must be ordered separately. For compatible screw caps, select either disposable phenolic caps with rubber liners or reusable screw caps with PTFE liners for improved chemical resistance.
Specifications
- Catalog No:
- 33502-138
- 33502-140
- 89090-066
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Certificación ambiental:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
Specifications
Screw Caps
Product Information
Frequently Bought Together