Orders LinkContactUsLinkComponent
Difco™ DRBC Agar, BD
Difco™ DRBC Agar, BD
Difco™ DRBC Agar, 500 g
Catalog # 90003-540
Número de proveedor #:  258710
Difco™ DRBC Agar, BD
Catalog # 90003-540
CAS Number:  
Difco™ DRBC Agar, BD
Difco™ DRBC Agar, 500 g
CAS Number:  
Price
$6,500.00
Log in Para disponibilidad
Each (500g)
Product Details & Documents
Difco™ DRBC Agar (Dichloran Rose Bengal Chloramphenicol) supports good growth of yeasts and molds while inhibiting bacteria and the spreading of rapidly growing molds.
Documents
Especificaciones
Especificaciones
  • Description:
    Difco™ DRBC Agar
  • Size:
    500 g