Difco™ DRBC Agar, BD
Difco™ DRBC Agar, 500 g
Supplier: BECTON DICKINSON DE MEXICO, S. MX
Número de proveedor #: 258710
Jump to:
Product Details & Documents
Difco™ DRBC Agar (Dichloran Rose Bengal Chloramphenicol) supports good growth of yeasts and molds while inhibiting bacteria and the spreading of rapidly growing molds.
Documents
Certificates
Buscar por número de loteEspecificaciones
Especificaciones
- Description:Difco™ DRBC Agar
- Size:500 g