Zobell Marine Agar and Broth, HiMedia Laboratories
Supplier: HIMEDIA LABORATORIES LLC
Certificates
About this item
Zobell Marine Agar is used in the cultivation of heterotrophic marine bacteria, while Zobell Marine Broth is used in the isolation and enumeration of heterotrophic marine bacteria.
Frequently Bought Together