ContactUsLinkComponent
Accessories for InLab® Expert Pro 3-in-1 pH Electrodes, Double Junction, Mettler Toledo
Accessories for InLab® Expert Pro 3-in-1 pH Electrodes, Double Junction, Mettler Toledo
Catalog # 11238-458
Product available on GSA Advantage®
undefined
Accessories for InLab® Expert Pro 3-in-1 pH Electrodes, Double Junction, Mettler Toledo
Catalog # 11238-458
Supplier Number:  51350076
Product available on GSA Advantage®
Specifications
  • Descripción:
    FRISCOLYT-B® Electrolytes for reference electrodes, 250 ml (for measurement at low temperature and for media with organic compounds [oil, proteins etc.])
  • Referencia:
    11238-458
  • Nº de proveedor:
    51350076